CAS RN 26186-00-5
CAS RN 26186-00-5
IUPAC Name:
heptadec-1-yne
Synonyms:
heptadec-1-yne
Molecular Weight (g/mol):
236.44
Molecular Formula:
C17H32
InChi Key:
DQDNKQWGBZFFRA-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC#C
1
–
1
of
1
results
1
1-Heptadecyne 97.0+%, TCI America™
CAS: 26186-00-5 Molecular Formula: C17H32 Molecular Weight (g/mol): 236.443 MDL Number: MFCD00015087 InChI Key: DQDNKQWGBZFFRA-UHFFFAOYSA-N Synonym: Pentadecylacetylene PubChem CID: 141274 IUPAC Name: heptadec-1-yne SMILES: CCCCCCCCCCCCCCCC#C