Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptadecyne 97.0+%, TCI America™
SDP

Catalog No. H04401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04401G Supplier TCI America Supplier No. H04401G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26186-00-5
Molecular Formula C17H32
Molecular Weight (g/mol) 236.443
MDL Number MFCD00015087
InChI Key DQDNKQWGBZFFRA-UHFFFAOYSA-N
Synonym Pentadecylacetylene
PubChem CID 141274
IUPAC Name heptadec-1-yne
SMILES CCCCCCCCCCCCCCCC#C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 236.44
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Heptadecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.