accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2ONH2CH3

CAS RN 2620-27-1

IUPAC Name: methyl({2-[2-(methylazaniumyl)ethoxy]ethyl})azanium
Synonyms: methyl({2-[2-(methylammonio)ethoxy]ethyl})azanium
Molecular Weight (g/mol): 134.22
Molecular Formula: C6H18N2O
InChi Key: VXPJBVRYAHYMNY-UHFFFAOYSA-P
SMILES: C[NH2+]CCOCC[NH2+]C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)