Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Bis(methylamino)-3-oxapentane, 98%, Thermo Scientific™

Catalog No. AC275860025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC275860025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC275860025 Supplier Thermo Scientific Chemicals Supplier No. 275860025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-27-1
Molecular Formula C6H16N2O
Molecular Weight (g/mol) 132.207
InChI Key VXPJBVRYAHYMNY-UHFFFAOYSA-N
Synonym 2,2'-oxybis n-methylethanamine, ethanamine,2,2'-oxybis n-methyl, ethanamine, 2,2'-oxybis n-methyl, 1,5-bis methylamino-3-oxapentane, methyl 2-2-methylamino ethoxy ethyl amine, bmaop, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
PubChem CID 2724770
IUPAC Name N-methyl-2-[2-(methylamino)ethoxy]ethanamine
SMILES CNCCOCCNC

Specifications

Density 0.8720g/mL
Boiling Point 76.0°C to 78.0°C (20.0 mmHg)
Refractive Index 1.4370 to 1.4390
Linear Formula CH3NHCH2CH2OCH2CH2NHCH3
Quantity 2.5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 132.21
Chemical Name or Material 1,5-Bis(methylamino)-3-oxapentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.