accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONBr

CAS RN 26218-75-7

IUPAC Name: methyl 6-bromopyridine-2-carboxylate
Synonyms: methyl 6-bromopyridine-2-carboxylate
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: SGNCOKUHMXLGAH-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Br)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results