accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2MgCl

CAS RN 2622-05-1

IUPAC Name: chloro(prop-2-en-1-yl)magnesium
Synonyms: chloro(prop-2-en-1-yl)magnesium
Molecular Weight (g/mol): 100.83
Molecular Formula: C3H5ClMg
InChi Key: PLYLAFITHJUEGX-UHFFFAOYSA-M
SMILES: Cl[Mg]CC=C
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)