Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allylmagnesium Chloride Solution

Catalog No. 225908800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-590-8800ML 800 mL
22-590-818L 18 L
22-590-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 22-590-8800ML Supplier Sigma Aldrich Supplier No. 225908800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5'sa-disubstituted hydantoins.

Specifications

CAS 2622-05-1
Density 0.995 g/mL at 25°C
Molecular Formula C3H5ClMg
Linear Formula CH2=CHCH2MgCl
MDL Number MFCD00000473
Quantity 800 mL
Molecular Weight (g/mol) 100.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.