accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)BrOHO

CAS RN 2623-86-1

IUPAC Name: (3S)-3-bromobutanoic acid
Synonyms: (S)-(+)-3-bromobutanoic acid
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: HAIUIAZIUDPZIE-VKHMYHEASA-N
SMILES: C[C@H](Br)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results