Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobutyric acid, 97%, Thermo Scientific™

Catalog No. AAB2568403
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2568403 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2568403 Supplier Thermo Scientific Chemicals Supplier No. B2568403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2623-86-1
Flash Point >110°C (230°F)
Odor Unpleasant
Molecular Formula C4H7BrO2
Quantity 1g
MDL Number MFCD00053318
UN Number UN3265
Synonym 3-bromobutyric acid, butanoic acid, 3-bromo, acmc-1cjov, #, 3-bromo butyric acid, 3-bromobutyric acid gc
InChI Key HAIUIAZIUDPZIE-UHFFFAOYSA-N
SMILES CC(CC(=O)O)Br
IUPAC Name 3-bromobutanoic acid
Molecular Weight (g/mol) 167.002
PubChem CID 102860
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material 3-Bromobutyric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.