accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2629-54-1

IUPAC Name: (2S)-2-azaniumyl-3-(3-fluorophenyl)propanoate
Synonyms: (2S)-2-ammonio-3-(3-fluorophenyl)propanoate
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: VWHRYODZTDMVSS-QMMMGPOBSA-N
SMILES: [NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)