Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-DL-phenylalanine, 98%

Catalog No. AAL0729706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0729706 5 g
AAL0729703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0729706 Supplier Thermo Scientific Chemicals Supplier No. L0729706
  • $194.00
    $164.65 / Each of 1

    Save $29.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2629-54-1 | C9H10FNO2 | 183.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2629-54-1
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00004273,MFCD00066450
InChI Key VWHRYODZTDMVSS-QMMMGPOBSA-N
Synonym 3-fluoro-dl-phenylalanine, 3-fluorophenylalanine, 2-amino-3-3-fluorophenyl propanoic acid, m-fluorophenylalanine, dl-3-fluorophenylalanine, h-dl-phe 3-f-oh, m-fluoro-dl-phenylalanine, alanine, 3-fluorophenyl, 3-fluoro-3-phenylalanine, dl-phenylalanine, 3-fluoro
PubChem CID 9976
IUPAC Name 2-amino-3-(3-fluorophenyl)propanoic acid
SMILES [NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O

Specifications

Melting Point ∽245°C (decomposition)
Beilstein 2939807
Quantity 5 g
Formula Weight 183.18
Percent Purity 98%
Chemical Name or Material 3-Fluoro-DL-phenylalanine
EINECSNumber 220-104-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.