accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3ONHOHN

CAS RN 26305-13-5

IUPAC Name: 5,6-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5,6-dimethyl-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: PZVLJGKJIMBYNP-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=O)NC(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results