accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFCH3H3CCH3NNH3CH3CCH3

CAS RN 263163-17-3

IUPAC Name: 1,3-di-tert-butyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1,3-di-tert-butylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 268.11
Molecular Formula: C11H21BF4N2
InChi Key: OOFLHRYFPBGTPQ-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC(C)(C)N1C=C[N+](=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results