Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di-tert-butylimidazolium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. D37115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37115G Supplier TCI America Supplier No. D37115G
Add to Cart
Add to Cart

Specifications

CAS 263163-17-3
Melting Point 210°C
Molecular Formula C11H21BF4N2
Quantity 5 g
MDL Number MFCD07784425
UN Number 1759
Synonym ItBu.HBF4
InChI Key OOFLHRYFPBGTPQ-UHFFFAOYSA-N
SMILES [B-](F)(F)(F)F.CC(C)(C)N1C=C[N+](=C1)C(C)(C)C
IUPAC Name 1,3-ditert-butylimidazol-1-ium;tetrafluoroborate
Molecular Weight (g/mol) 268.107
PubChem CID 16217979
Formula Weight 268.11
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di-tert-butylimidazolium Tetrafluoroborate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.