accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONH2N

CAS RN 263339-24-8

IUPAC Name: 4-[(4-aminophenyl)methyl]-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-[(4-aminophenyl)methyl]-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 240.32
Molecular Formula: C11H16N2O2S
InChi Key: TZMCILRCGMFRJC-UHFFFAOYSA-N
SMILES: NC1=CC=C(CN2CCS(=O)(=O)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results