accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 263369-88-6

IUPAC Name: 3,4-bis(2-methoxyphenyl)-1,2-dihydro-1,2-chloraphosphinin-1-uide
Synonyms: 3,4-bis(2-methoxyphenyl)-1,2-dihydro-1,2-chloraphosphinin-1-uide
Molecular Weight (g/mol): 331.76
Molecular Formula: C18H17ClO2P
InChi Key: WVGQIZOJKMMSSK-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=C(C=C[Cl-]P1)C1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results