Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobis(2-methoxyphenyl)phosphine, 98+%

Catalog No. AAH2589406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2589406 5 g
AAH2589403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2589406 Supplier Thermo Scientific Chemicals Supplier No. H2589406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 263369-88-6 | C18H17ClO2P- | 331.756 g/mol

Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 263369-88-6
Molecular Formula C18H17ClO2P-
Molecular Weight (g/mol) 331.756
MDL Number MFCD08064037
InChI Key WVGQIZOJKMMSSK-UHFFFAOYSA-N
Synonym bis 2-methoxyphenyl chlorphosphine, acmc-20alnz, phosphinous chloride,p,p-bis 2-methoxyphenyl
PubChem CID 57371085
SMILES COC1=CC=CC=C1C2=C(P[Cl-]C=C2)C3=CC=CC=C3OC

Specifications

Melting Point 111°C to 114°C
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 280.69
Percent Purity ≥98%
Chemical Name or Material Chlorobis(2-methoxyphenyl)phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.