accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2636-26-2

IUPAC Name: O-4-cyanophenyl O,O-dimethyl phosphorothioate
Synonyms: cyanox cynock sumitomo S 4084 ciafos epa pesticide chemical code 268200 cyanophos caswell no. 268A CYAP
Molecular Weight (g/mol): 243.22
Molecular Formula: C9H10NO3PS
InChi Key: SCKHCCSZFPSHGR-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results