accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOON

CAS RN 26371-07-3

IUPAC Name: 3-(piperidin-1-yl)propanoic acid
Synonyms: piperidinepropionic acid
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: LPDGWMLCUHULJF-UHFFFAOYSA-N
SMILES: OC(=O)CCN1CCCCC1
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)