accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 263744-72-5

IUPAC Name: [2-(2-ethyl-5-methoxy-1H-indol-3-yl)ethyl]dimethylamine
Synonyms: EMDT
Molecular Weight (g/mol): 246.35
Molecular Formula: C15H22N2O
InChi Key: ZEYRDXUWJDGTLD-UHFFFAOYSA-N
SMILES: CCC1=C(CCN(C)C)C2=CC(OC)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results