accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26394-17-2

IUPAC Name: cyclopentanesulfonyl chloride
Synonyms: cyclopentanesulfonyl chloride
Molecular Weight (g/mol): 168.64
Molecular Formula: C5H9ClO2S
InChi Key: HZVKYZHPDGEECE-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1CCCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)