Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanesulfonyl chloride, 90%

Catalog No. AC437830050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437830050 5 g Glass bottle
AC437830010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437830050 Supplier Thermo Scientific Chemicals Supplier No. 437830050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26394-17-2 | C5H9ClO2S | 168.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26394-17-2
Molecular Formula C5H9ClO2S
Molecular Weight (g/mol) 168.64
MDL Number MFCD04117774
InChI Key HZVKYZHPDGEECE-UHFFFAOYSA-N
Synonym cyclopentylsulfonyl chloride, cyclopentanesulphonyl chloride, chlorocyclopentylsulfone, cyclopentylsulfonylchloride, acmc-1ccr5, cyclopentanesulfonylchloride, cyclopentyl sulfonyl chloride, chlorosulphonyl cyclopentane, cyclopentane sulfonyl chloride, cyclopentane sulphonyl chloride
PubChem CID 3717197
IUPAC Name cyclopentanesulfonyl chloride
SMILES C1CCC(C1)S(=O)(=O)Cl

Specifications

Color Brown
Boiling Point 95°C to 98°C (2.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 168.64
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Cyclopentanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.