accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOH

CAS RN 26402-22-2

IUPAC Name: 2,3-dihydroxypropyl decanoate
Synonyms: glyceryl caprate
Molecular Weight (g/mol): 246.35
Molecular Formula: C13H26O4
InChi Key: LKUNXBRZDFMZOK-UHFFFAOYNA-N
SMILES: CCCCCCCCCC(=O)OCC(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results