Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocaprin 98.0+%, TCI America™
SDP

Catalog No. M10725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10725G 5 g
M10721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10725G Supplier TCI America Supplier No. M10725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26402-22-2
Molecular Formula C13H26O4
Molecular Weight (g/mol) 246.35
MDL Number MFCD00056656
InChI Key LKUNXBRZDFMZOK-UHFFFAOYNA-N
Synonym Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol
PubChem CID 92926
ChEBI CHEBI:75551
IUPAC Name 2,3-dihydroxypropyl decanoate
SMILES CCCCCCCCCC(=O)OCC(O)CO

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monocaprin
EINECSNumber (2)-0669
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.