accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2642-71-9

IUPAC Name: O,O-diethyl {[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)methyl]sulfanyl}phosphonothioate
Synonyms: bionex guthion (ethyl) gusation A ethyl gusathion caswell no. 344 ethyl guthion gusathion athyl-gusathion azinfos-ethyl ethyl homolog of guthion
Molecular Weight (g/mol): 345.37
Molecular Formula: C12H16N3O3PS2
InChi Key: RQVGAIADHNPSME-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCN1N=NC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results