accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOO

CAS RN 26444-49-5

IUPAC Name: 4-methylphenyl diphenyl phosphate
Synonyms: cresyl diphenyl phosphate
Molecular Weight (g/mol): 340.32
Molecular Formula: C19H17O4P
InChi Key: OJUZRFGUKHQNJX-UHFFFAOYSA-N
SMILES: CC1=CC=C(OP(=O)(OC2=CC=CC=C2)OC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)