Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl methylphenyl phosphate, mixture of isomers, 94%

Catalog No. AA42092AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42092AP 500 mL
AA42092AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42092AP Supplier Thermo Scientific Chemicals Supplier No. 042092AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26444-49-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26444-49-5
MDL Number MFCD00014920
Synonym Phosphoric acid diphenyl methylphenyl ester; Cresyl diphenyl phosphate

Specifications

Melting Point -38°C
Color Yellow
Density 1.21 g/mL
Boiling Point 390°C
Flash Point 232°C (450°F)
Refractive Index 1.563
Quantity 500 mL
UN Number UN3082
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Diphenyl methylphenyl phosphate, Mixture of isomers
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 247-693-8
RTECSNumber TC5520000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.