accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26455-31-2

IUPAC Name: 1-nitro-4-(propan-2-yloxy)benzene
Synonyms: P-nitrophenyl-I-propyl ether
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: GSOJVDNLACMALV-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results