Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropoxy-4-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. I057925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I057925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I057925G Supplier TCI America Supplier No. I057925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26455-31-2
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD01327671
InChI Key GSOJVDNLACMALV-UHFFFAOYSA-N
Synonym Isopropyl 4-Nitrophenyl Ether
PubChem CID 12625304
IUPAC Name 1-nitro-4-propan-2-yloxybenzene
SMILES CC(C)OC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 25 g
Formula Weight 181.19
Chemical Name or Material 1-Isopropoxy-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.