accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSSCH3NSOOCH3

CAS RN 2651-15-2

IUPAC Name: N-[bis(methylsulfanyl)methylidene]-4-methylbenzene-1-sulfonamide
Synonyms: N-[bis(methylsulfanyl)methylidene]-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 275.4
Molecular Formula: C10H13NO2S3
InChi Key: OWIPGZGAUSIOAX-UHFFFAOYSA-N
SMILES: CSC(SC)=NS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results