accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 2651-46-9

IUPAC Name: (4-chlorobutoxy)benzene
Synonyms: (4-chlorobutoxy)benzene
Molecular Weight (g/mol): 184.66
Molecular Formula: C10H13ClO
InChi Key: JKXCPAVECBFBOC-UHFFFAOYSA-N
SMILES: ClCCCCOC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results