Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxybutyl Chloride 95.0+%, TCI America™
SDP

Catalog No. P138625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P138625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P138625G Supplier TCI America Supplier No. P138625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2651-46-9
Molecular Formula C10H13ClO
Molecular Weight (g/mol) 184.663
MDL Number MFCD00001012
InChI Key JKXCPAVECBFBOC-UHFFFAOYSA-N
Synonym 4-Chlorobutyl Phenyl Ether
PubChem CID 75857
IUPAC Name 4-chlorobutoxybenzene
SMILES C1=CC=C(C=C1)OCCCCCl

Specifications

Boiling Point 144°C
Quantity 25 g
Formula Weight 184.66
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenoxybutyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.