CAS RN 26523-78-4
CAS RN 26523-78-4
IUPAC Name:
tris(4-nonylphenyl) phosphite
Synonyms:
tris(nonylphenyl)phosphite
Molecular Weight (g/mol):
689.02
Molecular Formula:
C45H69O3P
InChi Key:
MGMXGCZJYUCMGY-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC1=CC=C(OP(OC2=CC=C(CCCCCCCCC)C=C2)OC2=CC=C(CCCCCCCCC)C=C2)C=C1
1
–
2
of
2
results
1
Sigma Aldrich Tris(nonylphenyl) phosphite
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2
Tris(nonylphenyl)phosphite, SPEX CertiPrep™
CAS: 26523-78-4 Molecular Formula: C45H69O3P Molecular Weight (g/mol): 689.018 InChI Key: WGKLOLBTFWFKOD-UHFFFAOYSA-N PubChem CID: 85595 IUPAC Name: tris(2-nonylphenyl) phosphite SMILES: CCCCCCCCCC1=CC=CC=C1OP(OC2=CC=CC=C2CCCCCCCCC)OC3=CC=CC=C3CCCCCCCCC