Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(nonylphenyl) phosphite

Catalog No. 441805250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-180-5250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-180-5250ML Supplier Sigma Aldrich Supplier No. 441805250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(nonylphenyl) phosphite (TNPP) is an antioxidant that is majorly used as a stabilizer to improve the performance of polyethylene by decomposing the hydroperoxides. It can also be used to enhance the thermal stability by extending the polymeric chains.TNPP in combination with irganox is used to protect polyamide 6 (PA6) from oxidative degradation during the preparation of flame retardant nanocomposites. TNPP can be used as a stabilizer that prevents the molecular weight reduction and increases the tensile strength of the composite. It can also be used as a chain extender to improve the polymeric viscosity during the melt-blending of poly(hydroxybutyrate-co-hydroxyvalerate) based clay nanocomposites which find potential application as packaging materials.

Specifications

CAS 26523-78-4
Density 0.99 g/mL (at 25°C)
Boiling Point >360°C
Molecular Formula C45H69O3P
Refractive Index n20/D 1.528 (literature)
Linear Formula (C9H19C6H4O)3 P
Quantity 250 mL
MDL Number MFCD00015301
Synonym Tri(monononylphenyl) phosphite
Molecular Weight (g/mol) 689
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.