accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2653-64-7

IUPAC Name: (1Z)-1-[2-(naphthalen-1-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms: (1Z)-1-[2-(naphthalen-1-yl)hydrazin-1-ylidene]naphthalen-2-one
Molecular Weight (g/mol): 298.35
Molecular Formula: C20H14N2O
InChi Key: DJCJDQSTGATKMZ-XDOYNYLZSA-N
SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results