Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthylamine Bordeaux (diluted with Barium Sulfate) 90.0+%, TCI America™
SDP

Catalog No. N030725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N030725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N030725G Supplier TCI America Supplier No. N030725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2653-64-7
Molecular Formula C20H14N2O
Molecular Weight (g/mol) 298.345
MDL Number MFCD00071296
InChI Key DJCJDQSTGATKMZ-XDOYNYLZSA-N
PubChem CID 5351167
IUPAC Name (1Z)-1-(naphthalen-1-ylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC=C2NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Color Purple-Red
Quantity 25 g
Formula Weight 298.35
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Naphthylamine Bordeaux (diluted with Barium Sulfate)
EINECSNumber (5)-3230
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.