accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3CH3OOO(E)

CAS RN 26544-38-7

IUPAC Name: 3-[(2E)-4,6,8-trimethylnon-2-en-2-yl]oxolane-2,5-dione
Synonyms: 3-[(2E)-4,6,8-trimethylnon-2-en-2-yl]oxolane-2,5-dione
Molecular Weight (g/mol): 266.38
Molecular Formula: C16H26O3
InChi Key: WBRBLKRDTHYRRV-MDWZMJQENA-N
SMILES: CC(C)CC(C)CC(C)\C=C(/C)C1CC(=O)OC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)