Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dodecenylsuccinic anhydride, mixture of isomers

Catalog No. 4432711L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-327-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-327-11L Supplier Sigma Aldrich Supplier No. 4432711L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dodecenylsuccinic anhydride, mixture of isomers (DDSA) belongs to the class of long chained aliphatic anhydrides that are used as catalysts and organic acid anhydride based hardeners that find usage in electrical industry.DDSA can be used as a hardener that facilitates the curing the of epoxy resin. It can also be used in the formation of glycine based surfactants.

Specifications

CAS 26544-38-7
Density 1.005 g/mL (at 25°C)
Boiling Point 150°C (3 mmHg)
Molecular Formula C16H26O3
Refractive Index n20/D 1.479 (literature)
Linear Formula C16H26O3
Quantity 1 L
MDL Number MFCD00036197
Synonym DDSA
Molecular Weight (g/mol) 266.38
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.