accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH3CCH3NCH3CH3OCH3

CAS RN 26549-21-3

IUPAC Name: [4-(dimethylamino)-2,3-dimethoxybutyl]dimethylamine
Synonyms: [4-(dimethylamino)-2,3-dimethoxybutyl]dimethylamine
Molecular Weight (g/mol): 204.31
Molecular Formula: C10H24N2O2
InChi Key: VYQCQNCBTMHEFI-UHFFFAOYNA-N
SMILES: COC(CN(C)C)C(CN(C)C)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results