Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane 97.0+%, TCI America™
SDP

Catalog No. D23965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23965G 5 g
D23961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23965G Supplier TCI America Supplier No. D23965G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26549-21-3
Molecular Formula C10H24N2O2
Molecular Weight (g/mol) 204.314
MDL Number MFCD00063598
InChI Key VYQCQNCBTMHEFI-UWVGGRQHSA-N
PubChem CID 12349546
IUPAC Name (2S,3S)-2,3-dimethoxy-N,N,N',N'-tetramethylbutane-1,4-diamine
SMILES CN(C)CC(C(CN(C)C)OC)OC

Specifications

Color Yellow
Quantity 5 g
Synonym (S,S)-(+)-DDB, (S,S)-(+)-2,3-Dimethoxy-N,N,N′,N′-tetramethyl-1,4-butanediamine
Formula Weight 204.31
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material (S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.