accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2655-27-8

IUPAC Name: N-pentylaniline
Synonyms: N-pentylaniline
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: UMNSMBWAESLVOC-UHFFFAOYSA-N
SMILES: CCCCCNC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results