Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Pentylaniline 98.0+%, TCI America™
SDP

Catalog No. P06235ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06235ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06235ML Supplier TCI America Supplier No. P06235ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2655-27-8
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.26
MDL Number MFCD00059431
InChI Key UMNSMBWAESLVOC-UHFFFAOYSA-N
Synonym N-Amylaniline
PubChem CID 75865
IUPAC Name N-pentylaniline
SMILES CCCCCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 245°C
Quantity 5 mL
Formula Weight 163.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Pentylaniline
EINECSNumber (3)-0206
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.