accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3HO

CAS RN 26550-55-0

IUPAC Name: 1-methoxypropan-2-ol
Synonyms: methoxyisopropanol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: ARXJGSRGQADJSQ-UHFFFAOYNA-N
SMILES: COCC(C)O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)