CAS RN 26576-46-5
CAS RN 26576-46-5
IUPAC Name:
3-oxo-N-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-5-yl)butanamide
Synonyms:
3-oxo-N-(2-oxo-1,3-dihydro-1,3-benzodiazol-5-yl)butanamide
Molecular Weight (g/mol):
233.23
Molecular Formula:
C11H11N3O3
InChi Key:
VEMDQCGHZNXORX-UHFFFAOYSA-N
SMILES:
CC(=O)CC(=O)NC1=CC=C2NC(=O)NC2=C1
1
–
1
of
1
results
1
5-(Acetoacetamido)-2-benzimidazolinone 98.0+%, TCI America™
CAS: 26576-46-5 Molecular Formula: C11H11N3O3 MDL Number: MFCD00444508