accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26586-55-0

IUPAC Name: 1-[4-(4-methylpiperazin-1-yl)phenyl]ethan-1-one
Synonyms: 1-[4-(4-methylpiperazin-1-yl)phenyl]ethanone
Molecular Weight (g/mol): 218.3
Molecular Formula: C13H18N2O
InChi Key: IRIZGAMYKHTLKS-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=C(C=C1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results