Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methylpiperazino)acetophenone, 98%, Thermo Scientific™

Catalog No. AC152912500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152912500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC152912500 Supplier Thermo Scientific Chemicals Supplier No. 152912500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26586-55-0
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
MDL Number MFCD00023297
InChI Key IRIZGAMYKHTLKS-UHFFFAOYSA-N
Synonym 1-4-4-methylpiperazino phenyl-1-ethanone, 1-4-4-methylpiperazin-1-yl phenyl ethanone, p-4-methylpiperazino-acetophenone, acetophenone, 4'-4-methyl-1-piperazinyl, 1-4-4-methylpiperazin-1-yl phenyl ethan-1-one, ethanone, 1-4-4-methyl-1-piperazinyl phenyl, ethanone,1-4-4-methyl-1-piperazinyl phenyl
PubChem CID 33551
IUPAC Name 1-[4-(4-methylpiperazin-1-yl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2CCN(CC2)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 100°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 218.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methylpiperazino)acetophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 247-827-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.