accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNNHO

CAS RN 26661-13-2

IUPAC Name: N-(2-oxo-2,3-dihydropyrimidin-4-yl)benzamide
Synonyms: N-(2-oxo-3H-pyrimidin-4-yl)benzamide
Molecular Weight (g/mol): 215.21
Molecular Formula: C11H9N3O2
InChi Key: XBDUZBHKKUFFRH-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=NC(=O)N1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)