accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26693-55-0

IUPAC Name: (1R,2S)-2-benzamidocyclohexane-1-carboxylate
Synonyms: (1R,2S)-2-benzamidocyclohexane-1-carboxylate
Molecular Weight (g/mol): 246.29
Molecular Formula: C14H16NO3
InChi Key: PUANNVQABXUYKU-NEPJUHHUSA-M
SMILES: [O-]C(=O)[C@@H]1CCCC[C@@H]1NC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results