Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-cis-2-Benzamidocyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B11205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11205G Supplier TCI America Supplier No. B11205G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26693-55-0
Molecular Formula C14H16NO3
Molecular Weight (g/mol) 246.29
MDL Number MFCD00078309
InChI Key PUANNVQABXUYKU-NEPJUHHUSA-M
PubChem CID 1268226
IUPAC Name (1R,2S)-2-benzamidocyclohexane-1-carboxylate
SMILES [O-]C(=O)[C@@H]1CCCC[C@@H]1NC(=O)C1=CC=CC=C1

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 247.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-cis-2-Benzamidocyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.