accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2675-79-8

IUPAC Name: 5-bromo-1,2,3-trimethoxybenzene
Synonyms: 5-bromo-1,2,3-trimethoxybenzene
Molecular Weight (g/mol): 247.09
Molecular Formula: C9H11BrO3
InChi Key: XAOOZMATJDXDQJ-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(OC)=C1OC
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)